Phenylmagnesium bromide, 1.6M solution in CPME, AcroSeal™
Phenylmagnesium bromide, 1.6M solution in CPME, AcroSeal™
Phenylmagnesium bromide, 1.6M solution in CPME, AcroSeal™
Phenylmagnesium bromide, 1.6M solution in CPME, AcroSeal™
Thermo Scientific Chemicals
Phenylmagnesium bromide, 1.6M solution in CPME, AcroSeal™
CAS: 100-58-3 | C6H5BrMg | 181.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Catalog number FSA445961000
Price (MYR)
740.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (MYR)
740.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhenylmagnesium bromide
CAS100-58-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
CAS5614-37-9