Ammonium hydrogen sulfate, extra pure
Ammonium hydrogen sulfate, extra pure
Ammonium hydrogen sulfate, extra pure
Ammonium hydrogen sulfate, extra pure
Thermo Scientific Chemicals
Ammonium hydrogen sulfate, extra pure
CAS: 7803-63-6 | H5NO4S | 115.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number FSA445910010
Price (MYR)
677.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
677.00
EA
Add to cart
Specifications
Boiling Point350.0°C
Chemical Name or MaterialAmmonium hydrogen sulfate
Melting Point147.0°C
CAS7803-63-6
Health Hazard 1GHS Signal Word: Danger