1,1,1,3,3,3-Hexafluoro-2-propanol, 99%, for analysis
1,1,1,3,3,3-Hexafluoro-2-propanol, 99%, for analysis
1,1,1,3,3,3-Hexafluoro-2-propanol, 99%, for analysis
1,1,1,3,3,3-Hexafluoro-2-propanol, 99%, for analysis
Thermo Scientific Chemicals
1,1,1,3,3,3-Hexafluoro-2-propanol, 99%, for analysis
CAS: 920-66-1 | C3H2F6O | 168.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
10 mL
Packaging:
Glass bottle
Catalog number FSA445820500
Price (MYR)
1,583.00
EA
Add to cart
Quantity:
50 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,583.00
EA
Add to cart
Specifications
Chemical Name or Material1,1,1,3,3,3-Hexafluoro-2-propanol
CAS920-66-1
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for