p-Xylene, 99%, for HPLC
p-Xylene, 99%, for HPLC
p-Xylene, 99%, for HPLC
p-Xylene, 99%, for HPLC
Thermo Scientific Chemicals
p-Xylene, 99%, for HPLC
CAS: 106-42-3 | C8H10 | 106.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA445680025
Price (MYR)
1,186.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
1,186.00
EA
Add to cart
Specifications
Chemical Name or Materialp-Xylene
Melting Point13.0°C
Absorbance0.01 max. at 380nm, 0.01 max. at 400nm, 0.05 max. at 350nm, 0.1 max. at 320nm, 1 max. at 294nm
CAS106-42-3
Health Hazard 1Danger