Carbon disulfide, 99.9%, for HPLC
Carbon disulfide, 99.9%, for HPLC
Carbon disulfide, 99.9%, for HPLC
Carbon disulfide, 99.9%, for HPLC
Thermo Scientific Chemicals
Carbon disulfide, 99.9%, for HPLC
CAS: 75-15- | CS2 | 76.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA445660010
Price (MYR)
864.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
864.00
EA
Add to cart
Specifications
Chemical Name or MaterialCarbon disulfide
Melting Point-111.0°C
CAS75-15-
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
Highly fla