Chlorobenzene, 99.9%, for HPLC
Chlorobenzene, 99.9%, for HPLC
Chlorobenzene, 99.9%, for HPLC
Chlorobenzene, 99.9%, for HPLC
Thermo Scientific Chemicals
Chlorobenzene, 99.9%, for HPLC
CAS: 108-90-7 | C6H5Cl | 112.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
Glass bottle
Catalog number FSA445651000
Price (MYR)
391.00
EA
Add to cart
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
391.00
EA
Add to cart
Specifications
Chemical Name or MaterialChlorobenzene
Melting Point-45.0°C
CAS108-90-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Flammable liquid and vapor.