Boron trifluoride, 12% (1.5M) in methanol, AcroSeal™
Boron trifluoride, 12% (1.5M) in methanol, AcroSeal™
Boron trifluoride, 12% (1.5M) in methanol, AcroSeal™
Boron trifluoride, 12% (1.5M) in methanol, AcroSeal™
Thermo Scientific Chemicals
Boron trifluoride, 12% (1.5M) in methanol, AcroSeal™
CAS: 373-57-9 | BF3 | 67.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Catalog number FSA445278000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
800 mL
Request bulk or custom format
Chemical Identifiers
CAS373-57-9, 67-56-1
Specifications
Boiling Point65.0°C
Chemical Name or MaterialBoron trifluoride
Melting Point-98.0°C
CAS373-57-9
Health Hazard 1GHS Signal Word: Danger
View more
Lewis acid
RUO – Research Use Only