Acycloguanosine, 98%
Acycloguanosine, 98%
Acycloguanosine, 98%
Acycloguanosine, 98%
Thermo Scientific Chemicals
Acycloguanosine, 98%
CAS: 59277-89-3 | C8H11N5O3 | 225.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
Packaging:
Glass bottle
Catalog number FSA445240100
Price (MYR)
377.00
EA
Add to cart
Quantity:
10 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
377.00
EA
Add to cart
Specifications
Chemical Name or MaterialAcycloguanosine
CAS59277-89-3
Health Hazard 1Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Skin corrosion/irritation (category 2)
Health Hazard 2Causes serious eye irritation, May cause respiratory irritation, Causes skin irritation
Melting Point256°C to 260°C