Petroleum ether, technical, boiling range 100-140°C, Thermo Scientific Chemicals
Petroleum ether, technical, boiling range 100-140°C, Thermo Scientific Chemicals
Petroleum ether, technical, boiling range 100-140°C, Thermo Scientific Chemicals
Petroleum ether, technical, boiling range 100-140°C, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Petroleum ether, technical, boiling range 100-140°C, Thermo Scientific Chemicals
Have Questions?
Catalog NumberQuantityPackaging
FSA44436025025 LMetal drum
Catalog number FSA444360250
Price (MYR)
1,361.00
EA
Add to cart
Quantity:
25 L
Packaging:
Metal drum
Request bulk or custom format
Price (MYR)
1,361.00
EA
Add to cart
Chemical Identifiers
CAS64742-49-0, 111-65-9, 142-82-5, 108-87-2, 110-82-7, 73513-42-5
Specifications
Chemical Name or MaterialPetroleum ether
CAS64742-49-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May be fatal if swallowed and enters airways.
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness o
CAS111-65-9