Ethylene glycol, technical
Ethylene glycol, technical
Ethylene glycol, technical
Ethylene glycol, technical
Thermo Scientific Chemicals
Ethylene glycol, technical
CAS: 107-21-1 | C2H6O2 | 62.068 g/mol
Have Questions?
Catalog NumberQuantity
FSA4442300101 L
Catalog number FSA444230010
Price (MYR)
212.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
212.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthylene glycol
Melting Point-13.0°C
CAS107-21-1
Health Hazard 1Warning
Health Hazard 2GHS H StatementHarmful if swallowed.May cause damage to organs through prolonged or repeated exposure.