2-Butanone, technical
2-Butanone, technical
2-Butanone, technical
2-Butanone, technical
2-Butanone, technical
Thermo Scientific Chemicals
2-Butanone, technical
CAS: 78-93-3 | C4H8O | 72.11 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA4441700505 LPlastic Drum
Catalog number FSA444170050
Price (MYR)
425.00
EA
Add to cart
Quantity:
5 L
Packaging:
Plastic Drum
Request bulk or custom format
Price (MYR)
425.00
EA
Add to cart
Specifications
Chemical Name or Material2-Butanone
Melting Point-87.0°C
CAS78-93-3
Health Hazard 1GHS Signal Word: Danger
Density0.8060g/mL