Acetoacetamide, 97%
Acetoacetamide, 97%
Acetoacetamide, 97%
Thermo Scientific Chemicals
Acetoacetamide, 97%
CAS: 5977-14-0 | C4H7NO2 | 101.105 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA442541000
Price (MYR)
343.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
343.00
Each
Specifications
Chemical Name or MaterialAcetoacetamide
CAS5977-14-0
CAS Min %96.0
CAS Max %100.0
Melting Point-53°C to 56°C