Check Order Status
2,3,4-Trifluorobenzyl bromide, 97%, Thermo Scientific™
Element Search
Structure Search
2,3,4-Trifluorobenzyl bromide, 97%, Thermo Scientific™
Have Questions?
Change view
Quantity:
5 g
1 g
Catalog number
FSA442260050
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material
2, 3, 4-Trifluorobenzyl bromide
CAS
157911-55-2
CAS Min %
96.0
Health Hazard 1
GHS Signal Word:
Danger
CAS Max %
100.0