Check Order Status
6-chloronicotinoyl chloride, 97%, Thermo Scientific™
Element Search
Structure Search
6-chloronicotinoyl chloride, 97%, Thermo Scientific™
Have Questions?
Catalog Number
Quantity
Packaging
FSA442250250
25 g
Glass bottle
Catalog number
FSA442250250
Price (MYR)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material
6-chloronicotinoyl chloride
CAS
58757-38-3
CAS Min %
96.0
Health Hazard 1
GHS Signal Word:
Danger
CAS Max %
100.0