Sodium methanesulfonate, 99%
Sodium methanesulfonate, 99%
Sodium methanesulfonate, 99%
Sodium methanesulfonate, 99%
Thermo Scientific Chemicals
Sodium methanesulfonate, 99%
CAS: 2386-57-4 | CH4NaO3S | 118.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
Catalog number FSA442111000
Price (MYR)
600.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
600.00
EA
Add to cart
Specifications
Chemical Name or MaterialSodium methanesulfonate
CAS2386-57-4
Health Hazard 1Warning
Health Hazard 2GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Health Hazard 3GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.