1-Aminohydantoin hydrochloride, 98%
1-Aminohydantoin hydrochloride, 98%
1-Aminohydantoin hydrochloride, 98%
1-Aminohydantoin hydrochloride, 98%
Thermo Scientific Chemicals
1-Aminohydantoin hydrochloride, 98%
CAS: 2827-56-7 | C3H5N3O2·ClH | 151.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA442050050
Price (MYR)
425.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
425.00
EA
Add to cart
Specifications
Chemical Name or Material1-Aminohydantoin hydrochloride
CAS2827-56-7
CAS Min %97.5
CAS Max %100.0
Melting Point201.0°C to 205.0°C