BOC-L-Tyrosine methyl ester, 97%
BOC-L-Tyrosine methyl ester, 97%
BOC-L-Tyrosine methyl ester, 97%
BOC-L-Tyrosine methyl ester, 97%
Thermo Scientific Chemicals
BOC-L-Tyrosine methyl ester, 97%
CAS: 4326-36-7 | C15H21NO5 | 295.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA442020050
Price (MYR)
459.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
459.00
EA
Add to cart
Specifications
Chemical Name or MaterialBOC-L-Tyrosine methyl ester
CAS4326-36-7
Melting Point106.0°C to 107.0°C
ColorWhite to Yellow
Infrared SpectrumAuthentic