2-Amino-4-chloropyridine, 97%
2-Amino-4-chloropyridine, 97%
2-Amino-4-chloropyridine, 97%
2-Amino-4-chloropyridine, 97%
Thermo Scientific Chemicals
2-Amino-4-chloropyridine, 97%
CAS: 19798-80-2 | C5H6ClN2 | 129.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA441460050
Price (MYR)
543.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
543.00
EA
Add to cart
Specifications
Chemical Name or Material2-Amino-4-chloropyridine
CAS19798-80-2
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0