Palladium(II) acetate, 99.9%, (trace metal basis)
Palladium(II) acetate, 99.9%, (trace metal basis)
Palladium(II) acetate, 99.9%, (trace metal basis)
Palladium(II) acetate, 99.9%, (trace metal basis)
Thermo Scientific Chemicals
Palladium(II) acetate, 99.9%, (trace metal basis)
CAS: 3375-31-3 | C4H6O4Pd | 224.51 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA441390050
Price (MYR)
6,352.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
6,352.00
EA
Add to cart
Specifications
Chemical Name or MaterialPalladium(II) Acetate
Melting Point205.0°C
CAS3375-31-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes serious eye damage.
May cause long lasting harmful effects to aquatic life.
View more
For ultra trace analysis
RUO – Research Use Only