N-Methylcarbazole, 99%, Thermo Scientific Chemicals
N-Methylcarbazole, 99%, Thermo Scientific Chemicals
N-Methylcarbazole, 99%, Thermo Scientific Chemicals
N-Methylcarbazole, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
N-Methylcarbazole, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA441050050
Price (MYR)
752.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
752.00
EA
Add to cart
Specifications
Chemical Name or MaterialN-Methylcarbazole
CAS1484-12-4
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0