Xylene cyanol FF, ∼75%
Xylene cyanol FF, ∼75%
Xylene cyanol FF, ∼75%
Xylene cyanol FF, ∼75%
Thermo Scientific Chemicals
Xylene cyanol FF, ∼75%
CAS: 2650-17-1 | C25H27N2NaO6S2 | 538.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number FSA440700500
Price (MYR)
1,105.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,105.00
EA
Add to cart
Specifications
Chemical Name or MaterialXylene cyanol FF
CAS2650-17-1
Assay Percent Notes∽75%
Molecular FormulaC25H27N2NaO6S2
MDL NumberMFCD00019481