Propidium iodide, 95%
Propidium iodide, 95%
Propidium iodide, 95%
Propidium iodide, 95%
Thermo Scientific Chemicals
Propidium iodide, 95%
CAS: 25535-16-4 | C27H34I2N4 | 668.39 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA440301000100 mgGlass bottle
Catalog number FSA440301000
Price (MYR)
1,354.00
EA
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,354.00
EA
Add to cart
Specifications
Chemical Name or MaterialPropidium iodide
CAS25535-16-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing genetic defects.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately