Potassium sulfite, 90%, pure
Potassium sulfite, 90%, pure
Potassium sulfite, 90%, pure
Potassium sulfite, 90%, pure
Thermo Scientific Chemicals
Potassium sulfite, 90%, pure
CAS: 10117-38-1 | K2O3S | 158.26 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA440212500250 gGlass Bottle
Catalog number FSA440212500
Price (MYR)
258.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
258.00
EA
Add to cart
Specifications
Chemical Name or MaterialPotassium sulfite
CAS10117-38-1
Quantity250 g
EINECS Number233-321-1
Assay Percent Range88% min. (Iodimetry)