2-Fluorophenethyl bromide, 97%
2-Fluorophenethyl bromide, 97%
2-Fluorophenethyl bromide, 97%
2-Fluorophenethyl bromide, 97%
Thermo Scientific Chemicals
2-Fluorophenethyl bromide, 97%
CAS: 91319-54-9 | C8H8BrF | 203.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA440120050
Price (MYR)
1,251.00
EA
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
1,251.00
EA
Add to cart
Specifications
Chemical Name or Material2-Fluorophenethyl bromide
CAS91319-54-9
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0