N,N-Dimethyl-L-phenylalanine, 99%
N,N-Dimethyl-L-phenylalanine, 99%
N,N-Dimethyl-L-phenylalanine, 99%
N,N-Dimethyl-L-phenylalanine, 99%
Thermo Scientific Chemicals
N,N-Dimethyl-L-phenylalanine, 99%
CAS: 17469-89-5 | C11H15NO2 | 193.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA440030050
Price (MYR)
831.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
831.00
EA
Add to cart
Specifications
Chemical Name or MaterialN, N-Dimethyl-L-phenylalanine
CAS17469-89-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0