N-Phenyl-2-naphthylamine, 97%
N-Phenyl-2-naphthylamine, 97%
N-Phenyl-2-naphthylamine, 97%
N-Phenyl-2-naphthylamine, 97%
Thermo Scientific Chemicals
N-Phenyl-2-naphthylamine, 97%
CAS: 135-88-6 | C16H13N | 219.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA439911000
Price (MYR)
959.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
959.00
EA
Add to cart
Specifications
Chemical Name or MaterialN-Phenyl-2-naphthylamine
CAS135-88-6
Health Hazard 1GHS Signal Word: Warning
Melting Point105°C to 108°C
EINECS Number205-223-9