4-Bromophenethyl bromide, 96%
4-Bromophenethyl bromide, 96%
4-Bromophenethyl bromide, 96%
4-Bromophenethyl bromide, 96%
Thermo Scientific Chemicals
4-Bromophenethyl bromide, 96%
CAS: 1746-28-7 | C8H8Br2 | 263.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Catalog number FSA439530100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Bromophenethyl bromide
CAS1746-28-7
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0