Eriochrome™ Black T, ACS reagent, indicator grade
Eriochrome™ Black T, ACS reagent, indicator grade
Eriochrome™ Black T, ACS reagent, indicator grade
Eriochrome™ Black T, ACS reagent, indicator grade
Thermo Scientific Chemicals
Eriochrome™ Black T, ACS reagent, indicator grade
CAS: 1787-61-7 | C20H12N3NaO7S | 461.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA439142500
Price (MYR)
381.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
381.00
EA
Add to cart
Specifications
Chemical Name or MaterialEriochrome™ Black T
CAS1787-61-7
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
Health Hazard 3GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Clarity(0.1%, water) Passes Test
View more
Indicator
RUO – Research Use Only