Magnesium carbonate, for biochemistry, specified according to the requirements of USP
Magnesium carbonate, for biochemistry, specified according to the requirements of USP
Magnesium carbonate, for biochemistry, specified according to the requirements of USP
Magnesium carbonate, for biochemistry, specified according to the requirements of USP
Thermo Scientific Chemicals
Magnesium carbonate, for biochemistry, specified according to the requirements of USP
CAS: 546-93-0 | CMgO3 | 84.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA439132500
Price (MYR)
634.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
634.00
EA
Add to cart
Specifications
Chemical Name or MaterialMagnesium carbonate
CAS546-93-0
ColorWhite
Physical FormPowder
Quantity250 g
View more
For biochemistry
RUO – Research Use Only