1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, 97%
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, 97%
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, 97%
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, 97%
Thermo Scientific Chemicals
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, 97%
CAS: 1892-57-5 | C8H17N3 | 155.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Packaging:
Glass bottle
Catalog number FSA438840500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide
CAS1892-57-5
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0
View more
Coupling reagent
RUO – Research Use Only