N-Hydroxysulfosuccinimide sodium salt, 95%
N-Hydroxysulfosuccinimide sodium salt, 95%
N-Hydroxysulfosuccinimide sodium salt, 95%
N-Hydroxysulfosuccinimide sodium salt, 95%
Thermo Scientific Chemicals
N-Hydroxysulfosuccinimide sodium salt, 95%
Coupling reagent | CAS: 106627-54-7 | C4H4NNaO6S | 217.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number FSA438650050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialN-Hydroxysulfosuccinimide sodium salt
CAS106627-54-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0
View more
Coupling reagent
RUO – Research Use Only