Methyl tridecanoate, 97%
Methyl tridecanoate, 97%
Methyl tridecanoate, 97%
Methyl tridecanoate, 97%
Thermo Scientific Chemicals
Methyl tridecanoate, 97%
CAS: 1731-88-0 | C14H28O2 | 228.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
1 g
5 g
Catalog number FSA438200100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl tridecanoate
CAS1731-88-0
Melting Point7°C
ColorColorless
Density0.8600g/mL