2,6-Pyridinedicarboxaldehyde, 97%
2,6-Pyridinedicarboxaldehyde, 97%
2,6-Pyridinedicarboxaldehyde, 97%
2,6-Pyridinedicarboxaldehyde, 97%
Thermo Scientific Chemicals
2,6-Pyridinedicarboxaldehyde, 97%
CAS: 5431-44-7 | C7H5NO2 | 135.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number FSA437752500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 6-Pyridinedicarboxaldehyde
CAS5431-44-7
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0