4-Cyanophenylacetic acid, 97%
4-Cyanophenylacetic acid, 97%
4-Cyanophenylacetic acid, 97%
4-Cyanophenylacetic acid, 97%
Thermo Scientific Chemicals
4-Cyanophenylacetic acid, 97%
CAS: 5462-71-5 | C9H7NO2 | 161.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA437730010
Price (MYR)
1,020.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,020.00
EA
Add to cart
Specifications
Chemical Name or Material4-Cyanophenylacetic acid
CAS5462-71-5
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0