Hydrogen tetrachloroaurate hydrate, 99.999%, (trace metal basis)
Hydrogen tetrachloroaurate hydrate, 99.999%, (trace metal basis)
Hydrogen tetrachloroaurate hydrate, 99.999%, (trace metal basis)
Hydrogen tetrachloroaurate hydrate, 99.999%, (trace metal basis)
Thermo Scientific Chemicals
Hydrogen tetrachloroaurate hydrate, 99.999%, (trace metal basis)
CAS: 27988-77-8 | AuCl4H | 339.77 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number FSA437170010
Price (MYR)
2,217.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,217.00
EA
Add to cart
Specifications
Chemical Name or Materialhydrogen tetrachloroaurate hydrate
Melting Point30.0°C
CAS27988-77-8
ColorYellow to Orange
Health Hazard 1GHS Signal Word: Danger
View more
for ultra trace analysis
RUO – Research Use Only