Rhodium(III) chloride hydrate, 99.95%, (trace metal basis)
Rhodium(III) chloride hydrate, 99.95%, (trace metal basis)
Rhodium(III) chloride hydrate, 99.95%, (trace metal basis)
Rhodium(III) chloride hydrate, 99.95%, (trace metal basis)
Rhodium(III) chloride hydrate, 99.95%, (trace metal basis)
Thermo Scientific Chemicals
Rhodium(III) chloride hydrate, 99.95%, (trace metal basis)
CAS: 20765-98-4 | Cl3Rh | 209.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number FSA437092500
Price (MYR)
2,028.00
EA
Add to cart
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,028.00
EA
Add to cart
Specifications
Chemical Name or MaterialRhodium(III) chloride hydrate
Melting Point100.0°C
CAS20765-98-4
ColorWhite
Health Hazard 1GHS Signal Word: Danger
View more
for ultra trace analysis
RUO – Research Use Only