Gold(III) chloride, 99.99+%, (trace metal basis)
Gold(III) chloride, 99.99+%, (trace metal basis)
Gold(III) chloride, 99.99+%, (trace metal basis)
Gold(III) chloride, 99.99+%, (trace metal basis)
Thermo Scientific Chemicals
Gold(III) chloride, 99.99+%, (trace metal basis)
CAS: 13453-07-1 | AuCl3 | 303.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA437080010
Price (MYR)
1,756.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,756.00
EA
Add to cart
Specifications
Chemical Name or MaterialGold(III) chloride
Melting Point254.0°C
CAS13453-07-1
ColorBrown to Red
Health Hazard 1GHS Signal Word: Warning
View more
For ultra trace analysis
RUO – Research Use Only