Tetraammineplatinum(II) chloride hydrate, 99.995%, (trace metal basis)
Tetraammineplatinum(II) chloride hydrate, 99.995%, (trace metal basis)
Tetraammineplatinum(II) chloride hydrate, 99.995%, (trace metal basis)
Tetraammineplatinum(II) chloride hydrate, 99.995%, (trace metal basis)
Tetraammineplatinum(II) chloride hydrate, 99.995%, (trace metal basis)
Thermo Scientific Chemicals
Tetraammineplatinum(II) chloride hydrate, 99.995%, (trace metal basis)
CAS: 108374-32-9 | Cl2H12N4Pt | 334.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
5 g
Packaging:
Glass bottle
Catalog number FSA437005000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTetraammineplatinum(II) chloride hydrate
CAS108374-32-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a
View more
for ultra trace analysis
RUO – Research Use Only