Pyrazinecarbonitrile, 99%
Pyrazinecarbonitrile, 99%
Pyrazinecarbonitrile, 99%
Pyrazinecarbonitrile, 99%
Thermo Scientific Chemicals
Pyrazinecarbonitrile, 99%
CAS: 19847-12-2 | C5H3N3 | 105.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA436920100
Price (MYR)
284.00
EA
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
284.00
EA
Add to cart
Specifications
Chemical Name or MaterialPyrazinecarbonitrile
CAS19847-12-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.