Ammonium dihydrogen phosphate, 99.999%, (trace metal basis)
Ammonium dihydrogen phosphate, 99.999%, (trace metal basis)
Ammonium dihydrogen phosphate, 99.999%, (trace metal basis)
Ammonium dihydrogen phosphate, 99.999%, (trace metal basis)
Thermo Scientific Chemicals
Ammonium dihydrogen phosphate, 99.999%, (trace metal basis)
CAS: 7722-76-1 | H6NO4P | 115.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number FSA436791000
Price (MYR)
702.00
EA
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
702.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium dihydrogen phosphate
Melting Point190.0°C
CAS7722-76-1
ColorWhite or Colorless
Physical FormCrystals
View more
for ultra trace analysis
RUO – Research Use Only