Trioctylphosphine, 90%, technical grade, AcroSeal™
Trioctylphosphine, 90%, technical grade, AcroSeal™
Trioctylphosphine, 90%, technical grade, AcroSeal™
Trioctylphosphine, 90%, technical grade, AcroSeal™
Trioctylphosphine, 90%, technical grade, AcroSeal™
Thermo Scientific Chemicals
Trioctylphosphine, 90%, technical grade, AcroSeal™
CAS: 4731-53-7 | C24H51P | 370.64 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA436461000100 mLAcroSeal™ Glass Bottle
Catalog number FSA436461000
Price (MYR)
1,125.00
EA
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
1,125.00
EA
Add to cart
Specifications
Chemical Name or MaterialTrioctylphosphine
CAS4731-53-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 3GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.