2,4-Bis(trifluoromethyl)benzyl bromide, 95%
2,4-Bis(trifluoromethyl)benzyl bromide, 95%
2,4-Bis(trifluoromethyl)benzyl bromide, 95%
2,4-Bis(trifluoromethyl)benzyl bromide, 95%
2,4-Bis(trifluoromethyl)benzyl bromide, 95%
CAS: 140690-56-8 | C9H5BrF6 | 307.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number FSA436350050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material2, 4-Bis(trifluoromethyl)benzyl bromide
CAS140690-56-8
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0