Pyridoxamine dihydrochloride, 98%
Pyridoxamine dihydrochloride, 98%
Pyridoxamine dihydrochloride, 98%
Pyridoxamine dihydrochloride, 98%
Thermo Scientific Chemicals
Pyridoxamine dihydrochloride, 98%
CAS: 524-36-7 | C8H12N2O2·2ClH | 241.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA436250050
Price (MYR)
1,637.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,637.00
EA
Add to cart
Specifications
Chemical Name or MaterialPyridoxamine dihydrochloride
CAS524-36-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee