Check Order Status
2,3-Difluorophenylacetic acid, 97%, Thermo Scientific™
Element Search
Structure Search
2,3-Difluorophenylacetic acid, 97%, Thermo Scientific™
Have Questions?
Change view
Quantity:
1 g
Catalog number
FSA435470010
Price (MYR)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material
2, 3-Difluorophenylacetic acid
CAS
145689-41-4
CAS Min %
96.0
Health Hazard 1
GHS Signal Word:
Warning
CAS Max %
100.0