4-Bromo-3-methylbenzoic acid, 97%
4-Bromo-3-methylbenzoic acid, 97%
4-Bromo-3-methylbenzoic acid, 97%
4-Bromo-3-methylbenzoic acid, 97%
Thermo Scientific Chemicals
4-Bromo-3-methylbenzoic acid, 97%
CAS: 7697-28-1 | C8H6BrO2 | 214.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA434780250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Bromo-3-methylbenzoic acid
CAS7697-28-1
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0