Urea, 99+%, for biochemistry
Urea, 99+%, for biochemistry
Urea, 99+%, for biochemistry
Urea, 99+%, for biochemistry
Thermo Scientific Chemicals
Urea, 99+%, for biochemistry
CAS: 57-13-6 | CH4N2O | 60.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic Bottle
Catalog number FSA434720010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialUrea
Melting Point131.0°C to 135.0°C
Name NoteFor Biochemistry
CAS57-13-6