2-Aminoethanethiol, 95%
2-Aminoethanethiol, 95%
2-Aminoethanethiol, 95%
2-Aminoethanethiol, 95%
Thermo Scientific Chemicals
2-Aminoethanethiol, 95%
CAS: 60-23-1 | C2H7NS | 77.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA434710250
Price (MYR)
1,592.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,592.00
EA
Add to cart
Specifications
Chemical Name or Material2-Aminoethanethiol
CAS60-23-1
CAS Min %94.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0