Decane, 99%, anhydrous, AcroSeal™
Decane, 99%, anhydrous, AcroSeal™
Decane, 99%, anhydrous, AcroSeal™
Decane, 99%, anhydrous, AcroSeal™
Thermo Scientific Chemicals
Decane, 99%, anhydrous, AcroSeal™
CAS: 124-18-5 | C10H22 | 142.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number FSA434600010
Price (MYR)
2,188.00
EA
Add to cart
Quantity:
1 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (MYR)
2,188.00
EA
Add to cart
Specifications
Chemical Name or MaterialDecane
CAS124-18-5
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May be fatal if swallowed and enters airways.
Flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Health Hazard 3GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.