Water, ACS reagent
Water, ACS reagent
Water, ACS reagent
Thermo Scientific Chemicals
Water, ACS reagent
CAS: 7732-18-5 | H2O | 18.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 L
1 L
Packaging:
Plastic Bottle
Catalog number FSA434400050
Price (MYR)
1,265.00
EA
Add to cart
Quantity:
5 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
1,265.00
EA
Add to cart
Specifications
AbsorbancePasses Test, 0.01 max. at 200nm, 0.005 max. at 300nm, 0.01 max. at 210nm, 0.005 max. at 254nm
Chemical Name or MaterialWater
Name NoteFor analysis ACS
CAS7732-18-5