Ammonium Sulfate, 99%, extra pure
Ammonium Sulfate, 99%, extra pure
Ammonium Sulfate, 99%, extra pure
Ammonium Sulfate, 99%, extra pure
Thermo Scientific Chemicals
Ammonium Sulfate, 99%, extra pure
CAS: 7783-20-2 | H8N2O4S | 132.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Plastic bottle
Catalog number FSA434380010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium sulfate
Melting Point280.0°C
Name NoteExtra Pure
CAS7783-20-2